Gene Transcripts

Gene Symbol
Snapc2
Entrez Gene ID
304204
Gene Name
small nuclear RNA activating complex, polypeptide 2
Gene Aliases
Chromosome Location
Chr. 12: 2537291 - 2540541 on Build Rnor_6.0
UniGene ID
Rn.162080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013121.1 NP_001013139.1 4 697
XM_008768956.1 XP_008767178.1 4 800
GenBank mRNA BC079077.1 - 4 697
FQ214278.1 - 3 527

View Full Product Details