Gene Transcripts

Gene Symbol
Ntn5
Entrez Gene ID
308587
Gene Name
netrin 5
Gene Aliases
RGD1565197
Chromosome Location
Chr. 1: 101667693 - 101678112 on Build Rnor_6.0
UniGene ID
Rn.225009
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173526.1 NP_001166997.1 3 768
XM_008759380.1 XP_008757602.1 5 2078
XM_008759381.1 XP_008757603.1 6 1928

View Full Product Details