Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003620.3 | NP_003611.1 | 2 | 909 | |
GenBank mRNA | AK299773.1 | - | 3 | 618 | |
AK312902.1 | - | 2 | 899 | ||
BC016480.1 | - | 2 | 825 | 3923593 | |
BC033893.2 | - | 2 | 783 | 5265521 | |
BC042418.1 | - | 2 | 823 | 5167004 | |
BC060877.1 | - | 2 | 909 | 5260081 | |
BT009780.1 | - | 2 | 677 | ||
KU178420.1 | - | 2 | 677 | ||
U78305.1 | - | 2 | 880 |