Gene Transcripts
- Gene Symbol
- RGD1560585
- Entrez Gene ID
- 317597
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 144001221 - 144001727 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100364877
- Entrez Gene ID
- 100364877
- Gene Name
- high-mobility group box 4-like
- Gene Aliases
- Chromosome Location
- Chr. X: 158694295 - 158694801 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100909696
- Entrez Gene ID
- 100909696
- Gene Name
- high mobility group protein B4-like
- Gene Aliases
- Chromosome Location
- Chr. X: 141614539 - 141615045 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC103694517
- Entrez Gene ID
- 103694517
- Gene Name
- high mobility group protein B4-like
- Gene Aliases
- Chromosome Location
- Chr. X: 139329414 - 139330118 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC688702
- Entrez Gene ID
- 688702
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- RGD1560254
- Chromosome Location
- Chr. X: 141611303 - 141612088 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1561558
- Entrez Gene ID
- 317314
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 158261211 - 158261717 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_228669.5 |
XP_228669.3
|
|
117 |
|
- Gene Symbol
- RGD1566008
- Entrez Gene ID
- 317604
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 140379673 - 140380419 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1566225
- Entrez Gene ID
- 317584
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 137197396 - 137197902 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details