Gene Transcripts

Gene Symbol
Magix
Entrez Gene ID
317379
Gene Name
MAGI family member, X-linked
Gene Aliases
Chromosome Location
Chr. X: 15669191 - 15676051 on Build Rnor_6.0
UniGene ID
Rn.107219
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014109.1 NP_001014131.1 5 747
GenBank mRNA BC078960.1 - 5 747

View Full Product Details