Gene Transcripts

Gene Symbol
Hdx
Entrez Gene ID
317617
Gene Name
highly divergent homeobox
Gene Aliases
RGD1563666
Chromosome Location
Chr. X: 82847895 - 83151516 on Build Rnor_6.0
UniGene ID
Rn.220606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134568.1 NP_001128040.1 4 1388

View Full Product Details