Gene Transcripts

Gene Symbol
Hnrnpr
Entrez Gene ID
319110
Gene Name
heterogeneous nuclear ribonucleoprotein R
Gene Aliases
Hnrpr
Chromosome Location
Chr. 5: 154668194 - 154699967 on Build Rnor_6.0
UniGene ID
Rn.195396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175603.2 NP_783193.2 5, 6 588
GenBank mRNA AY184814.1 - 4, 5 502
BC093598.1 - 5, 6 588 7315896

View Full Product Details