Gene Transcripts

Gene Symbol
Naa20
Entrez Gene ID
362228
Gene Name
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Gene Aliases
Nat5
Chromosome Location
Chr. 3: 140106726 - 140125047 on Build Rnor_6.0
UniGene ID
Rn.30327
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108595.1 NP_001102065.1 4 291
XM_006235155.2 XP_006235217.1 4 260
GenBank mRNA BC167020.1 - 4 251
FQ234868.1 - 4 216

View Full Product Details