Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105668.1 | NP_001099138.1 | 3, 4 | 569 | |
XM_006236102.2 | XP_006236164.1 | 3, 4 | 845 | ||
XM_006236103.2 | XP_006236165.1 | 3, 4 | 874 | ||
GenBank mRNA | CV106654.1 | - | 370 | ||
DN932316.1 | - | 3, 4 | 593 |