Gene Transcripts

Gene Symbol
Pomgnt1
Entrez Gene ID
362567
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases
Chromosome Location
Chr. 5: 135007343 - 135017220 on Build Rnor_6.0
UniGene ID
Rn.162088
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007747.1 NP_001007748.1 20 1889
XM_006238683.2 XP_006238745.1 19 1996
GenBank mRNA BC083641.1 - 20 1889

View Full Product Details