Gene Transcripts

Gene Symbol
Stat6
Entrez Gene ID
362896
Gene Name
signal transducer and activator of transcription 6, interleukin-4 induced
Gene Aliases
Chromosome Location
Chr. 7: 70946228 - 70963542 on Build Rnor_6.0
UniGene ID
Rn.6880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044250.1 NP_001037715.1 16 1910
GenBank mRNA DQ462201.1 - 16 1910

View Full Product Details