Gene Transcripts

Gene Symbol
Mtss1
Entrez Gene ID
362918
Gene Name
metastasis suppressor 1
Gene Aliases
Chromosome Location
Chr. 7: 98820803 - 98960736 on Build Rnor_6.0
UniGene ID
Rn.38488
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130563.1 NP_001124035.1 12 1223

View Full Product Details