Gene Transcripts

Gene Symbol
Sp100
Entrez Gene ID
363269
Gene Name
SP100 nuclear antigen
Gene Aliases
Chromosome Location
Chr. 9: 92675103 - 92718489 on Build Rnor_6.0
UniGene ID
Rn.15232
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014220.1 NP_001014242.1 8, 9 918
GenBank mRNA BC083808.1 - 8, 9 918

View Full Product Details