Gene Transcripts

Gene Symbol
Gcat
Entrez Gene ID
366959
Gene Name
glycine C-acetyltransferase
Gene Aliases
Chromosome Location
Chr. 7: 120263068 - 120269365 on Build Rnor_6.0
UniGene ID
Rn.43940
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024277.1 NP_001019448.1 2, 3 324
GenBank mRNA BC092591.1 - 2, 3 324

View Full Product Details