Gene Transcripts
- Gene Symbol
- Olr1699
- Entrez Gene ID
- 406011
- Gene Name
- olfactory receptor 1699
- Gene Aliases
- SCR D-9
- Chromosome Location
- Chr. 20: 1079474 - 1080439 on Build Rnor_6.0
- UniGene ID
- Rn.142680
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100362856
- Entrez Gene ID
- 100362856
- Gene Name
- olfactory receptor-like protein-like
- Gene Aliases
- Chromosome Location
- Chr. 20: 969096 - 970060 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Olr1696
- Entrez Gene ID
- 294158
- Gene Name
- olfactory receptor 1696
- Gene Aliases
- SCR D-8
- Chromosome Location
- Chr. 20: 878460 - 879425 on Build Rnor_6.0
- UniGene ID
- Rn.107045
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_206850.1 |
NP_996732.1
|
1 |
428 |
|
View Full Product Details