Gene Transcripts

Gene Symbol
Rnase13
Entrez Gene ID
497194
Gene Name
ribonuclease, RNase A family, 13 (non-active)
Gene Aliases
Chromosome Location
Chr. 15: 28322904 - 28325870 on Build Rnor_6.0
UniGene ID
Rn.142435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012231.1 NP_001012231.1 1 106
XM_006251896.2 XP_006251958.1 2 2244

View Full Product Details