Gene Transcripts

Gene Symbol
LOC499565
Entrez Gene ID
499565
Gene Name
LRRGT00038
Gene Aliases
Chromosome Location
Chr. 2: 87957363 - 87970337 on Build Rnor_6.0
UniGene ID
Rn.116010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047946.1 NP_001041411.1 1, 2 134
GenBank mRNA AY383693.1 - 1, 2 134

View Full Product Details