Gene Transcripts

Gene Symbol
Thsd7a
Entrez Gene ID
500032
Gene Name
thrombospondin, type I, domain containing 7A
Gene Aliases
RGD1566201
Chromosome Location
Chr. 4: 38664066 - 39102807 on Build Rnor_6.0
UniGene ID
Rn.208326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191970.1 NP_001178899.1 19 3741

View Full Product Details