Gene Transcripts

Gene Symbol
Helb
Entrez Gene ID
500837
Gene Name
helicase (DNA) B
Gene Aliases
RGD1560103
Chromosome Location
Chr. 7: 64860641 - 64887737 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241426.2 XP_006241488.1 12 3092
XM_006241428.2 XP_006241490.1 11 2845

View Full Product Details