Gene Transcripts

Gene Symbol
Gpr115
Entrez Gene ID
501106
Gene Name
G protein-coupled receptor 115
Gene Aliases
RGD1565216
Chromosome Location
Chr. 9: 20951249 - 20982493 on Build Rnor_6.0
UniGene ID
Rn.15113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006244640.2 XP_006244702.1 5 663
GenBank mRNA FQ222190.1 - 5 663

View Full Product Details