Gene Transcripts
- Gene Symbol
- RGD1563917
- Entrez Gene ID
- 501176
- Gene Name
- similar to Nuclear autoantigen Sp-100 (Speckled 100 kDa)
- Gene Aliases
- Chromosome Location
- Chr. 9: 92753841 - 92775830 on Build Rnor_6.0
- UniGene ID
- Rn.163087
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC088179.1 |
- |
3 |
286 |
|
View Full Product Details