Gene Transcripts

Gene Symbol
Defb41
Entrez Gene ID
641650
Gene Name
defensin beta 41
Gene Aliases
Chromosome Location
Chr. 15: 46179787 - 46182308 on Build Rnor_6.0
UniGene ID
Rn.191485
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037526.1 NP_001032615.1 2 95
GenBank mRNA AY621368.1 - 2 95

View Full Product Details