Gene Transcripts

Gene Symbol
Cstf2
Entrez Gene ID
683927
Gene Name
cleavage stimulation factor, 3' pre-RNA subunit 2
Gene Aliases
Chromosome Location
Chr. X: 104882727 - 104909746 on Build Rnor_6.0
UniGene ID
Rn.170541
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131014.1 NP_001124486.1 6, 7 707
XM_008773440.1 XP_008771662.1 6, 7 812
GenBank mRNA BC168251.1 - 6, 7 707

View Full Product Details