Gene Transcripts

Gene Symbol
Foxk2
Entrez Gene ID
303753
Gene Name
forkhead box K2
Gene Aliases
Chromosome Location
Chr. 10: 110468893 - 110518854 on Build Rnor_6.0
UniGene ID
Rn.44253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247890.2 XP_006247952.1 8 1837
GenBank mRNA BC168919.1 - 7 1305

View Full Product Details