Gene Transcripts

Gene Symbol
LOC684444
Entrez Gene ID
684444
Gene Name
similar to HIStone family member (his-41)
Gene Aliases
Chromosome Location
Chr. 2: 29247748 - 29248255 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749172.3 XP_003749220.1 1 379

View Full Product Details