Gene Transcripts

Gene Symbol
Pcca
Entrez Gene ID
687008
Gene Name
propionyl CoA carboxylase, alpha polypeptide
Gene Aliases
Chromosome Location
Chr. 15: 108960509 - 109306879 on Build Rnor_6.0
UniGene ID
Rn.6033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019330.1 NP_062203.1 18, 19 1764
XM_008770973.1 XP_008769195.1 18, 19 1704
GenBank mRNA CO574538.1 - 4, 5 403

View Full Product Details