Gene Transcripts

Gene Symbol
Ccdc137
Entrez Gene ID
688298
Gene Name
coiled-coil domain containing 137
Gene Aliases
Chromosome Location
Chr. 10: 109630059 - 109636480 on Build Rnor_6.0
UniGene ID
Rn.216435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143896.1 NP_001137368.1 1, 2 163
XM_008768483.1 XP_008766705.1 1, 2 203
GenBank mRNA BC158793.1 - 1, 2 130

View Full Product Details