Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014877.3 | NP_055692.2 | 13 | 1515 | |
XM_005257888.4 | XP_005257945.1 | 13 | 1563 | ||
XM_005257889.4 | XP_005257946.1 | 13 | 1563 | ||
XM_006722214.3 | XP_006722277.1 | 13 | 1563 | ||
XM_006722215.3 | XP_006722278.1 | 11 | 1435 | ||
XM_006722216.3 | XP_006722279.1 | 2 | 265 | ||
XM_011525544.2 | XP_011523846.1 | 13 | 1634 | ||
XM_017025477.1 | XP_016880966.1 | 11 | 1435 | ||
XM_017025478.1 | XP_016880967.1 | 14 | 1697 | ||
GenBank mRNA | BC094881.1 | - | 11 | 1382 | |
BC130582.1 | - | 13 | 1495 | 40146613 | |
BC144083.1 | - | 13 | 1495 | 9052603 | |
D29677.2 | - | 12 | 1473 |