Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004122.2 | NP_004113.1 | 1 | 813 | |
NM_198407.2 | NP_940799.1 | 1 | 813 | ||
GenBank mRNA | AY429112.1 | - | 1 | 770 | |
BC069068.1 | - | 1 | 770 | 7216881 | |
BC069374.1 | - | 1 | 770 | 7262191 | |
BC113547.1 | - | 1 | 813 | 8322599 | |
U60179.1 | - | 1 | 770 | ||
U60181.1 | - | 1 | 770 |