Gene Transcripts
- Gene Symbol
- RETN
- Entrez Gene ID
- 56729
- Gene Name
- resistin
- Gene Aliases
- ADSF, FIZZ3, RETN1, RSTN, XCP1
- Chromosome Location
- Chr. 19: 7669086 - 7670454 on Build GRCh38
- UniGene ID
- Hs.283091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193374.1 |
NP_001180303.1
|
2, 3 |
151 |
|
NM_020415.3 |
NP_065148.1
|
2, 3 |
160 |
|
GenBank mRNA |
AF205952.1 |
- |
1, 2 |
138 |
|
AF290874.1 |
- |
2, 3 |
160 |
|
AF323081.1 |
- |
2, 3 |
160 |
|
AY207314.1 |
- |
2, 3 |
151 |
|
AY359066.1 |
- |
1, 2 |
138 |
|
BC069302.1 |
- |
2, 3 |
160 |
7262685 |
BC101554.1 |
- |
2, 3 |
143 |
8069060 |
BC101560.1 |
- |
2, 3 |
143 |
8069066 |
View Full Product Details