Gene Transcripts

Gene Symbol
METTL26
Entrez Gene ID
84326
Gene Name
methyltransferase like 26
Gene Aliases
C16orf13, JFP2
Chromosome Location
Chr. 16: 634427 - 636366 on Build GRCh38
UniGene ID
Hs.239500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040160.2 NP_001035250.1 5 606
NM_001040161.2 NP_001035251.1 3 383
NM_001040162.2 NP_001035252.1 4 443
NM_001040165.2 NP_001035255.1 4 546
NM_001288710.1 NP_001275639.1 5 614
NM_032366.4 NP_115742.3 6 674
NR_109976.1 5 618
NR_109977.1 5 511
NR_109978.1 6 781
NR_109979.1 4 451
XM_011522713.2 XP_011521015.1 6 873
XM_011522714.2 XP_011521016.1 5 698
GenBank mRNA AI146806.1 - 4 440
AK057077.1 - 3 1572
AK294482.1 - 6 610
AK295628.1 - 5 542
BC007207.1 - 3 365 4054799
BG612714.1 - 5 474
BI762987.1 - 6 650
BM827705.1 - 4 378
BQ921757.1 - 579
BU539308.1 - 4 399

View Full Product Details