Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005499.2 | NP_005490.1 | 10 | 959 | |
XM_005258404.3 | XP_005258461.2 | 11 | 1240 | ||
XM_006722962.2 | XP_006723025.1 | 9 | 850 | ||
XM_011526304.2 | XP_011524606.1 | 11 | 1240 | ||
XM_017026134.1 | XP_016881623.1 | 11 | 1240 | ||
GenBank mRNA | AF079566.1 | - | 10 | 979 | |
AF090384.1 | - | 10 | 925 | ||
AF110957.1 | - | 10 | 889 | ||
AK023120.1 | - | 10 | 960 | ||
AK024114.1 | - | 2 | 134 | ||
AK124730.1 | - | 10 | 1100 | ||
AK299514.1 | - | 7 | 743 | ||
AK315521.1 | - | 10 | 936 | ||
AL136905.1 | - | 10 | 914 | ||
BC003153.1 | - | 10 | 937 | 2905977 | |
BT009781.1 | - | 10 | 889 | ||
CR456756.1 | - | 10 | 889 | ||
U35832.1 | - | 10 | 914 |