Gene Transcripts

Gene Symbol
BAGE4
Entrez Gene ID
85317
Gene Name
B melanoma antigen family member 4
Gene Aliases
CT2.4, MLL3P
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.545789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181704.1 NP_859055.1 363
GenBank mRNA AF339515.1 - 363

View Full Product Details