Gene Transcripts
- Gene Symbol
- SRRM1
- Entrez Gene ID
- 10250
- Gene Name
- serine and arginine repetitive matrix 1
- Gene Aliases
- 160-KD, POP101, SRM160
- Chromosome Location
- Chr. 1: 24643012 - 24673281 on Build GRCh38
- UniGene ID
- Hs.18192
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303448.1 |
NP_001290377.1
|
2, 3 |
343 |
|
NM_001303449.1 |
NP_001290378.1
|
2, 3 |
272 |
|
NM_005839.3 |
NP_005830.2
|
2, 3 |
343 |
|
XM_005245717.2 |
XP_005245774.1
|
2, 3 |
253 |
|
XM_005245718.2 |
XP_005245775.1
|
2, 3 |
253 |
|
XM_005245720.2 |
XP_005245777.1
|
2, 3 |
253 |
|
XM_005245721.2 |
XP_005245778.1
|
2, 3 |
253 |
|
XM_005245722.2 |
XP_005245779.1
|
2, 3 |
253 |
|
XM_011540478.2 |
XP_011538780.1
|
2, 3 |
267 |
|
XM_011540479.1 |
XP_011538781.1
|
2, 3 |
434 |
|
XM_011540480.1 |
XP_011538782.1
|
2, 3 |
434 |
|
XM_011540481.1 |
XP_011538783.1
|
2, 3 |
434 |
|
XM_011540482.1 |
XP_011538784.1
|
2, 3 |
434 |
|
XM_011540483.1 |
XP_011538785.1
|
2, 3 |
434 |
|
XM_017000011.1 |
XP_016855500.1
|
2, 3 |
267 |
|
XM_017000012.1 |
XP_016855501.1
|
2, 3 |
267 |
|
XM_017000013.1 |
XP_016855502.1
|
2, 3 |
267 |
|
XM_017000014.1 |
XP_016855503.1
|
2, 3 |
267 |
|
XM_017000015.1 |
XP_016855504.1
|
2, 3 |
267 |
|
XM_017000016.1 |
XP_016855505.1
|
2, 3 |
289 |
|
XM_017000017.1 |
XP_016855506.1
|
2, 3 |
267 |
|
XM_017000018.1 |
XP_016855507.1
|
2, 3 |
253 |
|
XM_017000019.1 |
XP_016855508.1
|
2, 3 |
253 |
|
XM_017000020.1 |
XP_016855509.1
|
2, 3 |
253 |
|
XM_017000021.1 |
XP_016855510.1
|
2, 3 |
253 |
|
XM_017000022.1 |
XP_016855511.1
|
2, 3 |
253 |
|
XM_017000023.1 |
XP_016855512.1
|
2, 3 |
434 |
|
XM_017000024.1 |
XP_016855513.1
|
2, 3 |
434 |
|
XM_017000025.1 |
XP_016855514.1
|
2, 3 |
253 |
|
XM_017000026.1 |
XP_016855515.1
|
2, 3 |
253 |
|
XM_017000027.1 |
XP_016855516.1
|
2, 3 |
434 |
|
XM_017000028.1 |
XP_016855517.1
|
2, 3 |
434 |
|
XM_017000029.1 |
XP_016855518.1
|
2, 3 |
434 |
|
XM_017000030.1 |
XP_016855519.1
|
2, 3 |
434 |
|
XM_017000031.1 |
XP_016855520.1
|
2, 3 |
434 |
|
XM_017000032.1 |
XP_016855521.1
|
2, 3 |
434 |
|
XM_017000033.1 |
XP_016855522.1
|
2, 3 |
434 |
|
XM_017000034.1 |
XP_016855523.1
|
2, 3 |
267 |
|
GenBank mRNA |
AF048977.1 |
- |
2, 3 |
124 |
|
AF419855.1 |
- |
2, 3 |
346 |
|
AK302498.1 |
- |
2, 3 |
162 |
|
AK307419.1 |
- |
2, 3 |
272 |
|
BC036187.1 |
- |
2, 3 |
171 |
4827738 |
BG720378.1 |
- |
2, 3 |
346 |
|
DC353932.1 |
- |
2, 3 |
252 |
|
KU178512.1 |
- |
2, 3 |
119 |
|
KU178513.1 |
- |
2, 3 |
119 |
|
View Full Product Details