Gene Transcripts

Gene Symbol
NBPF1
Entrez Gene ID
55672
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
AB13, AB14, AB23, AD2, NBG, NBPF
Chromosome Location
Chr. 1: 16562427 - 16613605 on Build GRCh38
UniGene ID
Hs.467587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017940.4 NP_060410.2 2008
GenBank mRNA AY894575.1 - 13 1890

View Full Product Details