Gene Transcripts

Gene Symbol
Arhgap31
Entrez Gene ID
12549
Gene Name
Rho GTPase activating protein 31
Gene Aliases
5830477L08Rik, AU041750, Cdgap, D330026I07Rik
Chromosome Location
Chr. 16: 38598343 - 38713035 on Build GRCm38
UniGene ID
Mm.370848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020260.2 NP_064656.2 5, 6 901
GenBank mRNA AF151363.1 - 5, 6 542
AK018057.1 - 5, 6 767
AK138417.1 - 5, 6 1141
AK149975.1 - 5, 6 737
AK150109.1 - 5, 6 737
AK155729.1 - 5, 6 721
AK170685.1 - 5, 6 721
BC138870.1 - 5, 6 706 8861892

View Full Product Details