Gene Transcripts

Gene Symbol
Smim20
Entrez Gene ID
66278
Gene Name
small integral membrane protein 20
Gene Aliases
1110067B18Rik, 1810013D10Rik, N28078
Chromosome Location
Chr. 5: 53267106 - 53278540 on Build GRCm38
UniGene ID
Mm.27900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145433.1 NP_001138905.1 3 711
GenBank mRNA AK007474.1 - 4 682
BC030079.1 - 3 647

View Full Product Details