Gene Transcripts

Gene Symbol
Zcchc13
Entrez Gene ID
75064
Gene Name
zinc finger, CCHC domain containing 13
Gene Aliases
4930513O09Rik, Cnbp2
Chromosome Location
Chr. X: 103630586 - 103631664 on Build GRCm38
UniGene ID
Mm.159414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029158.2 NP_083434.1 1 357
GenBank mRNA AK015789.1 - 1 359
BC145772.1 - 1 346 40131075
BC145774.1 - 1 346 40131077

View Full Product Details