Gene Transcripts

Gene Symbol
EMILIN1
Entrez Gene ID
11117
Gene Name
elastin microfibril interfacer 1
Gene Aliases
EMI, EMILIN, gp115
Chromosome Location
Chr. 2: 27078567 - 27086403 on Build GRCh38
UniGene ID
Hs.63348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007046.3 NP_008977.1 7 3189
GenBank mRNA AF088916.1 - 7 2690
AL050138.1 - 7 2620
BC007530.1 - 8 1647
BC009947.2 - 4 1384
BC136279.1 - 7 2789 9020266
BC142638.1 - 7 1917
BC142688.1 - 7 3071 40148654

View Full Product Details