Gene Transcripts

Gene Symbol
RGD1304567
Entrez Gene ID
362671
Gene Name
similar to RIKEN cDNA A430005L14
Gene Aliases
Chromosome Location
Chr. 5: 171242899 - 171247344 on Build Rnor_6.0
UniGene ID
Rn.4079
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009711.1 NP_001009711.1 4 659
GenBank mRNA BC087640.1 - 4 659

View Full Product Details