Gene Transcripts

Gene Symbol
Cpeb4
Entrez Gene ID
303010
Gene Name
cytoplasmic polyadenylation element binding protein 4
Gene Aliases
Chromosome Location
Chr. 10: 15984520 - 16047209 on Build Rnor_6.0
UniGene ID
Rn.18066
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245978.2 XP_006246040.1 1 370
XM_006245979.2 XP_006246041.1 1 370
XM_006245980.2 XP_006246042.1 1 370

View Full Product Details