Gene Transcripts

Gene Symbol
Ralbp1
Entrez Gene ID
84014
Gene Name
ralA binding protein 1
Gene Aliases
Chromosome Location
Chr. 9: 113579107 - 113598477 on Build Rnor_6.0
UniGene ID
Rn.7107
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032067.1 NP_114456.1 2 787
GenBank mRNA BC089788.1 - 2 841
U28830.1 - 2 791
U82623.1 - 2 787

View Full Product Details