Gene Transcripts

Gene Symbol
RNF13
Entrez Gene ID
11342
Gene Name
ring finger protein 13
Gene Aliases
RZF
Chromosome Location
Chr. 3: 149812688 - 149962139 on Build GRCh38
UniGene ID
Hs.12333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007282.4 NP_009213.1 9 1416
NM_183381.2 NP_899237.1 8 1314
XM_005247092.3 XP_005247149.1 8 1273
XM_011512373.2 XP_011510675.1 10 1421
XM_011512374.2 XP_011510676.1 9 1319
XM_011512376.2 XP_011510678.1 8 724
XM_017005654.1 XP_016861143.1 9 1495
XM_017005655.1 XP_016861144.1 8 1396
XM_017005656.1 XP_016861145.1 9 1375
XM_017005657.1 XP_016861146.1 9 1508
XM_017005658.1 XP_016861147.1 8 1406
XM_017005659.1 XP_016861148.1 7 1143
XM_017005660.1 XP_016861149.1 7 1133
XM_017005661.1 XP_016861150.1 2 498
GenBank mRNA AF037204.1 - 8 783
AF070558.1 - 8 774
AK090638.1 - 8 1294
AK090771.1 - 5 403
AK313304.1 - 8 798
BC009781.1 - 8 791 3683407
BC009803.1 - 9 894 4274992
BC017878.1 - 8 756
BG546092.1 - 6 587
BM556246.1 - 7 669
CB109037.1 - 2 141
CR456804.1 - 7 632

View Full Product Details