Gene Transcripts
- Gene Symbol
- PLD6
- Entrez Gene ID
- 201164
- Gene Name
- phospholipase D family member 6
- Gene Aliases
- ZUC
- Chromosome Location
- Chr. 17: 17200990 - 17206332 on Build GRCh38
- UniGene ID
- Hs.729079
- Species
- Homo sapiens
- Gene Symbol
- FLCN
- Entrez Gene ID
- 201163
- Gene Name
- folliculin
- Gene Aliases
- BHD, FLCL
- Chromosome Location
- Chr. 17: 17206945 - 17237188 on Build GRCh38
- UniGene ID
- Hs.31652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309336.1 |
- |
12 |
1406 |
|
View Full Product Details