Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001713.1 | NP_001001713.1 | 7 | 751 | |
NM_001317740.1 | NP_001304669.1 | 6 | 911 | ||
NM_001317741.1 | NP_001304670.1 | 6 | 801 | ||
NM_001317742.1 | NP_001304671.1 | 7 | 720 | ||
NM_007341.2 | NP_031367.1 | 7 | 980 | ||
GenBank mRNA | BC006371.2 | - | 7 | 931 | |
BI819819.1 | - | 7 | 722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317744.1 | NP_001304673.1 | 9 | 1169 |