Gene Transcripts

Gene Symbol
UBTFL1
Entrez Gene ID
642623
Gene Name
upstream binding transcription factor, RNA polymerase I-like 1
Gene Aliases
C11orf27, HMGPI
Chromosome Location
Chr. 11: 90085950 - 90087131 on Build GRCh38
UniGene ID
Hs.719885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143975.1 NP_001137447.1 1 631

View Full Product Details