Gene Transcripts

Gene Symbol
KCNU1
Entrez Gene ID
157855
Gene Name
potassium calcium-activated channel subfamily U member 1
Gene Aliases
KCNMC1, KCa5, KCa5.1, Kcnma3, Slo3
Chromosome Location
Chr. 8: 36784324 - 36936125 on Build GRCh38
UniGene ID
Hs.13861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031836.2 NP_001027006.2 27 3237
NR_134267.1 28 3299
XM_011544424.1 XP_011542726.1 26 3103
XM_011544426.1 XP_011542728.1 26 3062
XM_017013144.1 XP_016868633.1 27 3091
GenBank mRNA AK302391.1 - 27 3188
BC028701.2 - 28 3240

View Full Product Details