Gene Transcripts

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
Chromosome Location
Chr. 2: 169527879 - 169573921 on Build GRCh38
UniGene ID
Hs.529276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281476.2 NP_001268405.1 14 2723
NM_001322046.1 NP_001308975.1 15 3525
NM_001322048.1 NP_001308977.1 14 2780
NM_001322049.1 NP_001308978.1 13 2654
NM_024622.5 NP_078898.3 15 2852
NR_104020.2 14 2719
XM_006712750.3 XP_006712813.1 14 2778
XM_006712751.3 XP_006712814.1 11 2863
XM_017004911.1 XP_016860400.1 15 4165
XM_017004912.1 XP_016860401.1 10 2734
GenBank mRNA AB058703.1 - 13 2124
AK025554.1 - 12 1964
AK055892.1 - 14 2634
AK074302.1 - 15 2743
AK130797.1 - 14 2643
AK225293.1 - 12 1964
AL832058.1 - 15 2735
BC032687.2 - 14 2181 5581443

View Full Product Details