Gene Transcripts

Gene Symbol
Avp
Entrez Gene ID
11998
Gene Name
arginine vasopressin
Gene Aliases
Vp, Vsp
Chromosome Location
Chr. 2: 130580677 - 130582541 on Build GRCm38
UniGene ID
Mm.6190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009732.1 NP_033862.1 1 106
GenBank mRNA AK138179.1 - 1 139
BC051997.1 - 1 119 5683720

View Full Product Details