Gene Transcripts

Gene Symbol
Gm11564
Entrez Gene ID
670496
Gene Name
predicted gene 11564
Gene Aliases
OTTMUSG00000002201
Chromosome Location
Chr. 11: 99814976 - 99815666 on Build GRCm38
UniGene ID
Mm.466614
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100614.1 NP_001094084.1 1 531

View Full Product Details